Assay Details
Target Gene Details
Entrez Gene ID: | 5586 |
Gene Name: | protein kinase N2 |
Gene Aliases: |
PAK2, PRK2, PRKCL2, PRO2042, Pak-2, STK7 |
Location: |
Chr.1:88684144-88836255 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKN2 | NM_001320707.1 | 1 | 110 | NP_001307636.1 |
| NM_001320708.1 | 1 | 110 | NP_001307637.1 | |
| NM_001320709.1 | 1 | 110 | NP_001307638.1 | |
| NM_006256.3 | 1 | 110 | NP_006247.1 | |
| XM_017001782.1 | 1 | 61 | XP_016857271.1 | |
| XM_017001783.1 | 1 | 61 | XP_016857272.1 | |
| AK298595.1 | ||||
| AK300304.1 | ||||
| DA451427.1 |
Target Gene Details
Entrez Gene ID: | 101927891 |
Gene Name: | PKN2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:88537513-88685204 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKN2-AS1 | NR_110682.1 | |||
| AK123834.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546773 | Chr.1:88320368 - 88696449 on Build GRCh38 | Loss |
|
| esv3586738 | Chr.1:88623180 - 88685035 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map