Assay Details
Target Gene Details
Entrez Gene ID: | 1269 |
Gene Name: | cannabinoid receptor 2 |
Gene Aliases: |
CB-2, CB2, CX5 |
Location: |
Chr.1:23870526-23913327 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNR2 | NM_001841.2 | NP_001832.1 | ||
| XM_011540629.2 | XP_011538931.1 | |||
| XM_017000261.1 | XP_016855750.1 | |||
| AJ430063.1 | CAD22548.1 | |||
| AM156854.1 | CAJ42137.1 | |||
| AM156855.1 | CAJ42138.1 | |||
| AM156856.1 | CAJ42139.1 | |||
| BC069722.1 | AAH69722.1 | |||
| BC074767.2 | AAH74767.1 | |||
| BC095545.1 | AAH95545.1 | |||
| EU517121.1 | ||||
| X74328.1 | CAA52376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893081 | Chr.1:23843968 - 23913320 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map