Assay Details
Target Gene Details
Entrez Gene ID: | 7737 |
Gene Name: | ring finger protein 113A |
Gene Aliases: |
Cwc24, RNF113, TTD5, ZNF183 |
Location: |
Chr.X:119870532-119871828 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF113A | NM_006978.2 | 1 | 364 | NP_008909.1 |
| AK314778.1 | 1 | 310 | ||
| AW001915.1 | 1 | 364 | ||
| BC000832.1 | 1 | 207 | AAH00832.1 | |
| BC020556.1 | 1 | 250 | AAH20556.1 | |
| BC128440.1 | 1 | 249 | ||
| BT007175.1 | 1 | 149 | AAP35839.1 | |
| X98253.1 | 1 | 363 | CAA66907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7057 | Chr.X:119862293 - 119894925 on Build GRCh38 | Insertion |
|
| esv2758888 | Chr.X:119823476 - 120195131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map