Assay Details
Target Gene Details
Entrez Gene ID: | 6166 |
Gene Name: | ribosomal protein L36a like |
Gene Aliases: |
RPL36A |
Location: |
Chr.14:49618688-49620685 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL36AL | NM_001001.4 | NP_000992.1 | ||
| AB063609.1 | BAC19836.1 | |||
| AK311959.1 | ||||
| BC000741.2 | AAH00741.1 | |||
| BC003145.1 | AAH03145.1 | |||
| BC070207.1 | AAH70207.1 | |||
| BC107716.1 | AAI07717.1 | |||
| BM973694.1 | ||||
| DB447621.1 | ||||
| HY121971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv976338 | Chr.14:49618712 - 49621719 on Build GRCh38 | Duplication |
|
| esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs139622801] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map