Assay Details
Target Gene Details
Entrez Gene ID: | 5261 |
Gene Name: | phosphorylase kinase catalytic subunit gamma 2 |
Gene Aliases: |
GSD9C |
Location: |
Chr.16:30748299-30761176 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHKG2 | NM_000294.2 | NP_000285.1 | ||
| NM_001172432.1 | NP_001165903.1 | |||
| AK289492.1 | ||||
| AK293551.1 | ||||
| BC002541.2 | AAH02541.1 | |||
| M31606.1 | AAA36442.1 | |||
| S73483.1 | AAD14119.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469724 | Chr.16:30621124 - 30828942 on Build GRCh38 | Loss |
|
| nsv833188 | Chr.16:30621125 - 30828942 on Build GRCh38 | Loss |
|
| esv2714189 | Chr.16:30748966 - 30749248 on Build GRCh38 | Deletion |
|
| nsv1118349 | Chr.16:30748479 - 30749279 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map