Assay Details
Target Gene Details
Entrez Gene ID: | 8715 |
Gene Name: | nucleolar protein 4 |
Gene Aliases: |
CT125, HRIHFB2255, NOLP |
Location: |
Chr.18:33851100-34224834 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOL4 | NM_001198546.1 | 10 | 2322 | NP_001185475.1 |
| NM_001198547.1 | 11 | 2403 | NP_001185476.1 | |
| NM_001198548.1 | 9 | 2208 | NP_001185477.1 | |
| NM_001198549.1 | 7 | 1618 | NP_001185478.1 | |
| NM_001282527.1 | 11 | 2252 | NP_001269456.1 | |
| NM_003787.4 | 11 | 2514 | NP_003778.2 | |
| XM_006722563.2 | 12 | 3271 | XP_006722626.1 | |
| XM_006722564.2 | 10 | 2064 | XP_006722627.1 | |
| XM_006722565.1 | 12 | 2290 | XP_006722628.1 | |
| XM_011526237.1 | 12 | 3268 | XP_011524539.1 | |
| XM_011526238.1 | 11 | 2880 | XP_011524540.1 | |
| XM_011526239.2 | 10 | 2272 | XP_011524541.1 | |
| XM_017026052.1 | 10 | 2102 | XP_016881541.1 | |
| XM_017026053.1 | 11 | 2254 | XP_016881542.1 | |
| XM_017026054.1 | 12 | 2382 | XP_016881543.1 | |
| XM_017026055.1 | 11 | 2729 | XP_016881544.1 | |
| XM_017026056.1 | 11 | 1983 | XP_016881545.1 | |
| XM_017026057.1 | 12 | 2846 | XP_016881546.1 | |
| XM_017026058.1 | 11 | 1993 | XP_016881547.1 | |
| XM_017026059.1 | 12 | 3861 | XP_016881548.1 | |
| XM_017026060.1 | 6 | 1440 | XP_016881549.1 | |
| AA430033.1 | 1 | 222 | ||
| AB017800.1 | 11 | 2524 | BAA34576.1 | |
| AK091463.1 | 11 | 2101 | ||
| AK296539.1 | 7 | 1601 | ||
| BC000313.2 | 9 | 1894 | AAH00313.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2668525 | Chr.18:33248704 - 33887857 on Build GRCh38 | Deletion |
|
| nsv458048 | Chr.18:33840046 - 33917382 on Build GRCh38 | Loss |
|
| nsv576643 | Chr.18:33671257 - 34001283 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map