Assay Details
Target Gene Details
Entrez Gene ID: | 375759 |
Gene Name: | chromosome 9 open reading frame 50 |
Gene Aliases: |
- |
Location: |
Chr.9:129612225-129622275 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C9orf50 | NM_199350.3 | NP_955382.3 | ||
| XM_011518655.1 | XP_011516957.1 | |||
| XM_011518656.1 | XP_011516958.1 | |||
| XM_011518657.2 | XP_011516959.1 | |||
| XM_011518658.2 | XP_011516960.1 | |||
| XM_011518659.1 | XP_011516961.1 | |||
| XM_011518660.1 | XP_011516962.1 | |||
| XM_011518661.2 | XP_011516963.1 | |||
| XM_011518662.2 | XP_011516964.1 | |||
| XM_011518663.1 | XP_011516965.1 | |||
| XM_017014708.1 | XP_016870197.1 | |||
| AK093122.1 | ||||
| AW958564.1 | ||||
| AW958565.1 | ||||
| BC112353.1 | AAI12354.1 | |||
| JF442034.1 |
Target Gene Details
Entrez Gene ID: | 28989 |
Gene Name: | N-terminal Xaa-Pro-Lys N-methyltransferase 1 |
Gene Aliases: |
AD-003, C9orf32, HOMT1A, METTL11A, NRMT, NTM1A |
Location: |
Chr.9:129608884-129636742 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTMT1 | NM_001286796.1 | NP_001273725.1 | ||
| BG468453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469901 | Chr.9:129597897 - 129681820 on Build GRCh38 | Loss |
|
| nsv1046477 | Chr.9:129608081 - 129997996 on Build GRCh38 | Gain |
|
| nsv951783 | Chr.9:129584522 - 129613921 on Build GRCh38 | Deletion |
|
| nsv831731 | Chr.9:129514951 - 129634646 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map