Assay Details
Target Gene Details
Entrez Gene ID: | 6692 |
Gene Name: | serine peptidase inhibitor, Kunitz type 1 |
Gene Aliases: |
HAI, HAI1, MANSC2 |
Location: |
Chr.15:40844018-40857655 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPINT1 | NM_001032367.1 | 10 | 1741 | NP_001027539.1 |
| NM_003710.3 | 11 | 1835 | NP_003701.1 | |
| NM_181642.2 | 11 | 1883 | NP_857593.1 | |
| XM_006720657.1 | 11 | 1846 | XP_006720720.1 | |
| XM_011521957.1 | 10 | 1787 | XP_011520259.1 | |
| XM_011521958.1 | 10 | 1739 | XP_011520260.1 | |
| AB000095.1 | 11 | 1806 | BAA25014.1 | |
| AK300195.1 | 12 | 1784 | ||
| AY296715.1 | 11 | 1766 | AAP44001.1 | |
| AY358969.1 | 11 | 1866 | AAQ89328.1 | |
| BC004140.1 | 10 | 1683 | AAH04140.1 | |
| BC018702.1 | 11 | 1827 | AAH18702.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054764 | Chr.15:40776427 - 40904212 on Build GRCh38 | Gain |
|
| nsv569228 | Chr.15:40814287 - 40901953 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs148433906] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map