Assay Details
Target Gene Details
Entrez Gene ID: | 1945 |
Gene Name: | ephrin A4 |
Gene Aliases: |
EFL4, EPLG4, LERK4 |
Location: |
Chr.1:155063737-155069553 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EFNA4 | NM_005227.2 | NP_005218.1 | ||
| NM_182689.1 | NP_872631.1 | |||
| NM_182690.2 | NP_872632.2 | |||
| AI459151.1 | ||||
| AI625552.1 | ||||
| AJ006352.1 | CAA06992.1 | |||
| AJ006353.1 | CAA06993.1 | |||
| BC107483.1 | AAI07484.1 | |||
| BE780161.1 | ||||
| BM740848.1 | ||||
| BM747625.1 | ||||
| BQ940212.1 | ||||
| CD671399.1 | ||||
| CR533569.1 | CAG38600.1 | |||
| U14188.1 | AAC50079.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
| esv3587616 | Chr.1:155055926 - 155098657 on Build GRCh38 | Gain |
|
| esv3587615 | Chr.1:155047853 - 155175013 on Build GRCh38 | Gain |
|
| nsv952212 | Chr.1:154998225 - 155087624 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map