Assay Details
Target Gene Details
Entrez Gene ID: | 54716 |
Gene Name: | solute carrier family 6 member 20 |
Gene Aliases: |
SIT1, XT3, Xtrp3 |
Location: |
Chr.3:45755449-45796553 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A20 | NM_020208.3 | 6 | 997 | NP_064593.1 |
| NM_022405.3 | 5 | 886 | NP_071800.1 | |
| XM_011533847.2 | 5 | 1311 | XP_011532149.1 | |
| XM_011533848.2 | 6 | 1007 | XP_011532150.1 | |
| AF075260.1 | 6 | 875 | AAC27755.1 | |
| AF125107.1 | 7 | 1061 | AAL75944.1 | |
| AJ276207.1 | 6 | 924 | CAB99310.1 | |
| AJ276208.1 | 5 | 813 | CAB99311.1 | |
| AL389979.1 | 1 | 1061 | CAB97535.1 | |
| BC126197.1 | 6 | 937 | ||
| BC136431.1 | 6 | 937 | ||
| BC144087.1 | 6 | 937 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834678 | Chr.3:45762802 - 45968764 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map