Assay Details
Target Gene Details
Entrez Gene ID: | 2553 |
Gene Name: | GA binding protein transcription factor beta subunit 1 |
Gene Aliases: |
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4TF1B, GABPB, GABPB-1, GABPB2, NRF2B1, NRF2B2 |
Location: |
Chr.15:50277192-50355429 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GABPB1 | NM_001320910.1 | 8 | 1327 | NP_001307839.1 |
| NM_001320915.1 | 8 | 1324 | NP_001307844.1 | |
| NM_002041.4 | 7 | 1282 | NP_002032.2 | |
| NM_005254.5 | 7 | 1282 | NP_005245.2 | |
| NM_016654.4 | 7 | 1246 | NP_057738.1 | |
| NM_016655.4 | 7 | 1246 | NP_057739.1 | |
| NM_181427.3 | 8 | 1270 | NP_852092.1 | |
| XM_005254274.3 | 7 | 990 | XP_005254331.1 | |
| XM_011521426.2 | 7 | 1243 | XP_011519728.1 | |
| XM_017022053.1 | 8 | 1025 | XP_016877542.1 | |
| XM_017022054.1 | 8 | 983 | XP_016877543.1 | |
| AK301350.1 | 7 | 1072 | ||
| AK303901.1 | 6 | 890 | ||
| BC016910.1 | 7 | 967 | AAH16910.1 | |
| BC036080.1 | 7 | 1040 | AAH36080.1 | |
| BC050702.1 | 7 | 987 | AAH50702.1 | |
| BQ573816.1 | 2 | 378 | ||
| BT006652.1 | 6 | 858 | AAP35298.1 | |
| D13316.1 | 8 | 960 | BAA02573.1 | |
| D13317.1 | 7 | 1027 | BAA02574.1 | |
| EU159454.1 | 6 | 860 | ||
| JQ313901.1 | 6 | 819 | ||
| U13045.1 | 7 | 1027 | AAA65707.1 | |
| U13046.1 | 7 | 991 | AAA65708.1 | |
| U13047.1 | 7 | 1027 | AAA65709.1 | |
| U13048.1 | 7 | 991 | AAA65710.1 |
More Information
Set Membership: |
|

Genomic Map