Assay Details
Target Gene Details
Entrez Gene ID: | 5921 |
Gene Name: | RAS p21 protein activator 1 |
Gene Aliases: |
CM-AVM, CMAVM, GAP, PKWS, RASA, RASGAP, p120, p120GAP, p120RASGAP |
Location: |
Chr.5:87267801-87391926 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASA1 | NM_002890.2 | 1 | 189 | NP_002881.1 |
| XM_011543525.1 | 1 | 641 | XP_011541827.1 | |
| XM_011543527.2 | 1 | 641 | XP_011541829.1 | |
| AB209106.1 | 1 | 189 | BAD92343.1 | |
| BC033015.2 | 1 | 177 | AAH33015.1 | |
| BC131816.1 | 1 | 433 | ||
| CR749722.1 | 1 | 554 | CAH18488.2 | |
| M23379.1 | 1 | 108 | AAA52517.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830379 | Chr.5:87234875 - 87411690 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map