Assay Details
Target Gene Details
Entrez Gene ID: | 80760 |
Gene Name: | inter-alpha-trypsin inhibitor heavy chain family member 5 |
Gene Aliases: |
ITI-HC5, PP14776 |
Location: |
Chr.10:7559270-7666998 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITIH5 | NM_001001851.2 | NP_001001851.1 | ||
| NM_030569.6 | NP_085046.5 | |||
| NM_032817.5 | NP_116206.4 | |||
| XM_011519713.2 | XP_011518015.1 | |||
| XM_011519714.2 | XP_011518016.1 | |||
| AB075833.1 | BAB85539.1 | |||
| AF318347.1 | AAL55854.1 | |||
| AK027375.1 | BAB55070.1 | |||
| AK027831.1 | BAB55397.1 | |||
| AK027849.1 | BAB55409.1 | |||
| AK075381.1 | ||||
| AL833203.1 | ||||
| AY238437.1 | AAO49812.1 | |||
| AY358426.1 | AAQ88792.1 | |||
| CR627109.1 | CAH10363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038398 | Chr.10:7501987 - 7614812 on Build GRCh38 | Gain |
|
| nsv1038131 | Chr.10:7416409 - 7706490 on Build GRCh38 | Gain |
|
| esv2759732 | Chr.10:7381534 - 7695776 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map