Assay Details
Target Gene Details
Entrez Gene ID: | 6361 |
Gene Name: | C-C motif chemokine ligand 17 |
Gene Aliases: |
A-152E5.3, ABCD-2, SCYA17, TARC |
Location: |
Chr.16:57396076-57416063 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL17 | NM_002987.2 | NP_002978.1 | ||
| XM_011523256.2 | XP_011521558.1 | |||
| XM_017023530.1 | XP_016879019.1 | |||
| BC069107.1 | AAH69107.1 | |||
| D43767.1 | BAA07824.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs371376805] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map