Assay Details
Target Gene Details
Entrez Gene ID: | 4595 |
Gene Name: | mutY DNA glycosylase |
Gene Aliases: |
MYH |
Location: |
Chr.1:45329242-45340925 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Exon 3 |
Target Gene Details
Entrez Gene ID: | 114034 |
Gene Name: | target of EGR1, member 1 (nuclear) |
Gene Aliases: |
hCaf1z |
Location: |
Chr.1:45339457-45343978 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOE1 | NM_025077.3 | 1 | 287 | NP_079353.3 |
| XM_005270412.3 | 1 | 252 | XP_005270469.1 | |
| XM_005270413.4 | 1 | 309 | XP_005270470.1 | |
| XM_011540569.2 | 1 | 500 | XP_011538871.1 | |
| AY169960.1 | 1 | 287 | AAN75441.1 | |
| DA887065.1 | 1 | 213 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2750837 | Chr.1:45330443 - 45442591 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs185172591] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map