Assay Details
Target Gene Details
Entrez Gene ID: | 10745 |
Gene Name: | putative homeodomain transcription factor 1 |
Gene Aliases: |
PHTF |
Location: |
Chr.1:113697202-113759888 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHTF1 | NM_001323041.1 | NP_001309970.1 | ||
| NM_001323042.1 | NP_001309971.1 | |||
| NM_001323043.1 | NP_001309972.1 | |||
| NM_001323044.1 | NP_001309973.1 | |||
| NM_001323045.1 | NP_001309974.1 | |||
| NM_001323046.1 | NP_001309975.1 | |||
| NM_001323047.1 | NP_001309976.1 | |||
| NM_001323048.1 | NP_001309977.1 | |||
| NM_001323049.1 | NP_001309978.1 | |||
| NM_001323050.1 | NP_001309979.1 | |||
| NM_006608.2 | NP_006599.2 | |||
| NR_136564.1 | ||||
| NR_136565.1 | ||||
| NR_136566.1 | ||||
| NR_136567.1 | ||||
| NR_136568.1 | ||||
| AJ011863.1 | CAB51572.1 | |||
| AK126773.1 | ||||
| BC002447.2 | AAH02447.1 | |||
| DA728266.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587245 | Chr.1:113699035 - 113723249 on Build GRCh38 | Loss |
|
| esv3587240 | Chr.1:113533246 - 114158910 on Build GRCh38 | Loss |
|
| esv3587239 | Chr.1:113510852 - 113866662 on Build GRCh38 | Loss |
|
| esv23869 | Chr.1:113320330 - 114358495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map