Assay Details
Target Gene Details
Entrez Gene ID: | 80304 |
Gene Name: | WD repeat and coiled coil containing |
Gene Aliases: |
C2orf44, PP384 |
Location: |
Chr.2:24029336-24047426 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDCP | NM_001142319.1 | 3 | 2345 | NP_001135791.1 |
| NM_025203.2 | 4 | 2463 | NP_079479.1 | |
| AF218013.1 | 3 | 909 | AAG17255.1 | |
| AK025598.1 | 4 | 2390 | BAB15186.1 | |
| BC035698.2 | 3 | 2280 | AAH35698.1 | |
| CX756689.1 | 3 | 671 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997445 | Chr.2:23357900 - 24374347 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map