Assay Details
Target Gene Details
Entrez Gene ID: | 29116 |
Gene Name: | myosin regulatory light chain interacting protein |
Gene Aliases: |
IDOL, MIR |
Location: |
Chr.6:16129086-16151015 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYLIP | NM_013262.3 | 3 | 639 | NP_037394.2 |
| XM_005249032.2 | 3 | 639 | XP_005249089.1 | |
| XM_005249033.2 | 2 | 363 | XP_005249090.1 | |
| XM_017010789.1 | 3 | 639 | XP_016866278.1 | |
| AF006003.1 | 3 | 552 | AAQ13408.1 | |
| AF187016.2 | 3 | 576 | AAF18974.1 | |
| AF212221.1 | 2 | 363 | AAF87323.1 | |
| AF258586.1 | 3 | 616 | AAG23789.1 | |
| AK074391.1 | 3 | 639 | ||
| AK314648.1 | 3 | 639 | ||
| BC002860.2 | 3 | 620 | AAH02860.1 | |
| BT007055.1 | 3 | 402 | AAP35704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950132 | Chr.6:16128770 - 16148769 on Build GRCh38 | Duplication |
|
| nsv5209 | Chr.6:16117893 - 16151799 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map