Assay Details
Target Gene Details
Entrez Gene ID: | 5320 |
Gene Name: | phospholipase A2 group IIA |
Gene Aliases: |
MOM1, PLA2, PLA2B, PLA2L, PLA2S, PLAS1, sPLA2 |
Location: |
Chr.1:19975431-19980439 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G2A | NM_000300.3 | 6 | 831 | NP_000291.1 |
| NM_001161727.1 | 6 | 800 | NP_001155199.1 | |
| NM_001161728.1 | 5 | 720 | NP_001155200.1 | |
| NM_001161729.1 | 5 | 754 | NP_001155201.1 | |
| AK291302.1 | 5 | 681 | ||
| BC005919.1 | 6 | 808 | AAH05919.1 | |
| BU626291.1 | 1 | 187 | ||
| M22430.1 | 5 | 671 | AAA36550.1 |
More Information
Set Membership: |
|

Genomic Map