Assay Details
Target Gene Details
Entrez Gene ID: | 219865 |
Gene Name: | olfactory receptor family 8 subfamily G member 5 |
Gene Aliases: |
OR11-298, OR8G5P, OR8G6 |
Location: |
Chr.11:124241158-124323283 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR8G5 | NM_001005198.1 | 1 | 152 | NP_001005198.1 |
| XM_011542662.2 | XP_011540964.1 | |||
| XM_011542663.2 | XP_011540965.1 | |||
| XM_011542664.2 | XP_011540966.1 | |||
| XM_011542665.2 | XP_011540967.1 | |||
| XM_011542666.2 | XP_011540968.1 | |||
| XM_011542667.2 | XP_011540969.1 | |||
| BC136820.1 | 1 | 175 | ||
| BC136822.1 | 1 | 175 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745184 | Chr.11:124250355 - 124265734 on Build GRCh38 | Deletion |
|
| nsv975993 | Chr.11:124264837 - 124265945 on Build GRCh38 | Duplication |
|
| esv2745180 | Chr.11:124207805 - 124265663 on Build GRCh38 | Deletion |
|
| dgv82n21 | Chr.11:123901052 - 124289279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map