Assay Details
Target Gene Details
Entrez Gene ID: | 80025 |
Gene Name: | pantothenate kinase 2 |
Gene Aliases: |
C20orf48, HARP, HSS, NBIA1, PKAN |
Location: |
Chr.20:3888839-3929882 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PANK2 | NM_001324191.1 | 1 | 243 | NP_001311120.1 |
| NM_001324192.1 | 1 | 243 | NP_001311121.1 | |
| NM_024960.5 | NP_079236.3 | |||
| NM_153638.3 | 1 | 243 | NP_705902.2 | |
| NR_136715.1 | 1 | 243 | ||
| XM_005260836.4 | XP_005260893.3 | |||
| XM_011529364.2 | 1 | 99 | XP_011527666.1 | |
| XM_011529365.2 | 1 | 99 | XP_011527667.1 | |
| XM_017028079.1 | XP_016883568.1 | |||
| AF494409.1 | 1 | 82 | AAN32907.1 | |
| AK021791.1 | BAB13897.1 | |||
| AL519408.3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645053 | Chr.20:3849918 - 3987827 on Build GRCh38 | Loss |
|
| nsv828673 | Chr.20:3738382 - 4184811 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map