Assay Details
Target Gene Details
Entrez Gene ID: | 203100 |
Gene Name: | HtrA serine peptidase 4 |
Gene Aliases: |
- |
Location: |
Chr.8:38974149-38988662 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTRA4 | NM_153692.3 | NP_710159.1 | ||
| XM_011544431.2 | XP_011542733.1 | |||
| AK075205.1 | BAC11470.1 | |||
| AK300197.1 | ||||
| BC057765.1 | AAH57765.1 |
More Information
Set Membership: |
|

Genomic Map