Assay Details
Target Gene Details
Entrez Gene ID: | 201973 |
Gene Name: | primase and DNA directed polymerase |
Gene Aliases: |
CCDC111, MYP22 |
Location: |
Chr.4:184649613-184694959 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763423 | Chr.4:184642465 - 184688651 on Build GRCh38 | Loss |
|
| nsv830168 | Chr.4:184626297 - 184801578 on Build GRCh38 | Loss |
|
| nsv1026074 | Chr.4:184384963 - 184780663 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74696256] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map