Assay Details
Target Gene Details
Entrez Gene ID: | 339983 |
Gene Name: | N-acetyltransferase 8 like |
Gene Aliases: |
CML3, NACED, NAT8-LIKE |
Location: |
Chr.4:2059512-2069089 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAT8L | NM_178557.3 | 3 | 2458 | NP_848652.2 |
| AK094797.1 | 3 | 2110 | BAC04426.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| dgv8909n54 | Chr.4:2058874 - 2075739 on Build GRCh38 | Loss |
|
| nsv949757 | Chr.4:2056974 - 2082173 on Build GRCh38 | Deletion |
|
| nsv822441 | Chr.4:1991240 - 2080440 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map