Assay Details
Target Gene Details
Entrez Gene ID: | 10486 |
Gene Name: | CAP, adenylate cyclase-associated protein, 2 (yeast) |
Gene Aliases: |
- |
Location: |
Chr.6:17393216-17557792 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAP2 | NM_006366.2 | 1 | 119 | NP_006357.1 |
| XM_011514233.2 | 1 | 408 | XP_011512535.1 | |
| AK295600.1 | ||||
| AK312362.1 | 1 | 246 | ||
| AL832154.1 | CAI46134.1 | |||
| BI667914.1 | 1 | 180 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5921n100 | Chr.6:17384160 - 17459117 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map