Assay Details
Target Gene Details
Entrez Gene ID: | 85455 |
Gene Name: | dispatched RND transporter family member 2 |
Gene Aliases: |
DISPB, HsT16908, LINC00594 |
Location: |
Chr.15:40354359-40378755 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DISP2 | NM_033510.2 | NP_277045.1 | ||
| XM_011522125.2 | XP_011520427.1 | |||
| XM_017022687.1 | XP_016878176.1 | |||
| AB051529.2 | ||||
| BC151833.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510660 | Chr.15:40260705 - 40429684 on Build GRCh38 | Deletion |
|
| esv34145 | Chr.15:40257416 - 40597626 on Build GRCh38 | Loss |
|
| nsv1160287 | Chr.15:40338635 - 40365794 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map