Assay Details
Target Gene Details
Entrez Gene ID: | 116369 |
Gene Name: | solute carrier family 26 member 8 |
Gene Aliases: |
SPGF3, TAT1 |
Location: |
Chr.6:35943514-36024855 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC26A8 | NM_001193476.1 | 20 | 2964 | NP_001180405.1 |
| NM_052961.3 | 20 | 2988 | NP_443193.1 | |
| NM_138718.2 | 18 | 2711 | NP_619732.2 | |
| XM_011514294.2 | 19 | 3129 | XP_011512596.1 | |
| XM_017010235.1 | 20 | 3014 | XP_016865724.1 | |
| AF314959.1 | 19 | 2858 | AAL26868.1 | |
| AF331522.1 | 20 | 2932 | AAK95666.1 | |
| AI015633.1 | ||||
| AK055314.1 | 10 | 2434 | ||
| AK057276.1 | 10 | 1839 | BAB71408.1 | |
| AK097410.1 | 14 | 2050 | ||
| AK122928.1 | 20 | 2964 | ||
| BC025408.2 | 18 | 2704 | AAH25408.1 |
More Information
Set Membership: |
|

Genomic Map