Assay Details
Target Gene Details
Entrez Gene ID: | 100873065 |
Gene Name: | PTCHD1 antisense RNA (head to head) |
Gene Aliases: |
DDX53-AS1, PTCHD1AS1, PTCHD1AS2 |
Location: |
Chr.X:22193005-23293146 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTCHD1-AS | NR_073010.2 | |||
| DA390816.1 |
Target Gene Details
Entrez Gene ID: | 158506 |
Gene Name: | zinc finger protein 645 |
Gene Aliases: |
CT138, HAKAIL |
Location: |
Chr.X:22272913-22274461 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF645 | NM_152577.3 | 1 | 604 | NP_689790.1 |
| AK098601.1 | 1 | 576 | BAC05348.1 | |
| BC074910.2 | 1 | 546 | AAH74910.1 | |
| BC126190.1 | 1 | 546 | ||
| BC126192.1 | 1 | 546 | ||
| GQ355336.1 | 1 | 604 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521384 | Chr.X:22198650 - 22953327 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map