Assay Details
Target Gene Details
Entrez Gene ID: | 23133 |
Gene Name: | PHD finger protein 8 |
Gene Aliases: |
JHDM1F, MRXSSD, ZNF422 |
Location: |
Chr.X:53936680-54048935 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHF8 | NM_001184896.1 | 22 | 3922 | NP_001171825.1 |
| NM_001184897.1 | NP_001171826.1 | |||
| NM_015107.2 | 22 | 4073 | NP_055922.1 | |
| XM_005261996.1 | XP_005262053.1 | |||
| XM_005261997.3 | XP_005262054.1 | |||
| XM_005261999.1 | XP_005262056.1 | |||
| XM_005262000.1 | XP_005262057.1 | |||
| XM_011530778.1 | XP_011529080.1 | |||
| XM_017029361.1 | XP_016884850.1 | |||
| XM_017029362.1 | 22 | 4321 | XP_016884851.1 | |
| AB029034.1 | 22 | 3749 | ||
| AK022788.1 | 10 | 2241 | ||
| AK027229.1 | 1 | 418 | ||
| AK304272.1 | ||||
| BC053861.1 | 22 | 3777 | AAH53861.1 | |
| CR933612.1 | 22 | 3908 | CAI45929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv524220 | Chr.X:53755908 - 53949955 on Build GRCh38 | Gain |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map