Assay Details
Target Gene Details
Entrez Gene ID: | 23452 |
Gene Name: | angiopoietin like 2 |
Gene Aliases: |
ARP2, HARP |
Location: |
Chr.9:127087349-127122803 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGPTL2 | NM_012098.2 | 5 | 2814 | NP_036230.1 |
| AF007150.1 | 1 | 671 | ||
| AK074726.1 | 4 | 1818 | BAC11164.1 | |
| AK075026.1 | 4 | 1616 | BAC11358.1 |
Target Gene Details
Entrez Gene ID: | 9649 |
Gene Name: | Ral GEF with PH domain and SH3 binding motif 1 |
Gene Aliases: |
RALGEF2, RALGPS1A |
Location: |
Chr.9:126914774-127223166 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
More Information
Set Membership: |
|

Genomic Map