Assay Details
Target Gene Details
Entrez Gene ID: | 347902 |
Gene Name: | adhesion molecule with Ig like domain 2 |
Gene Aliases: |
ALI1, AMIGO-2, DEGA |
Location: |
Chr.12:47075707-47080826 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMIGO2 | NM_001143668.1 | 3 | 2481 | NP_001137140.1 |
| NM_181847.4 | 2 | 2379 | NP_862830.1 | |
| XM_005268837.3 | 3 | 3361 | XP_005268894.1 | |
| AB079074.1 | 1 | 1110 | BAC81188.1 | |
| AB080610.1 | 1 | 1889 | BAC81189.1 | |
| AK091761.1 | ||||
| AK093111.1 | 1 | 1319 | ||
| AL833007.1 | 3 | 2177 | CAH56293.1 | |
| AY454159.1 | 2 | 2294 | AAR83271.1 | |
| BC047595.1 | 2 | 2294 | AAH47595.1 | |
| BC095477.1 | 3 | 2481 | AAH95477.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv558784 | Chr.12:46981125 - 47281897 on Build GRCh38 | Gain |
|
| nsv694 | Chr.12:47066933 - 47079599 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map