Assay Details
Target Gene Details
Entrez Gene ID: | 51451 |
Gene Name: | leucine carboxyl methyltransferase 1 |
Gene Aliases: |
CGI-68, LCMT, PPMT1 |
Location: |
Chr.16:25111709-25178231 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCMT1 | NM_001032391.1 | 3 | 416 | NP_001027563.1 |
| NM_016309.2 | 3 | 416 | NP_057393.2 | |
| XM_005255354.3 | 3 | 205 | XP_005255411.1 | |
| XM_011545862.2 | 4 | 530 | XP_011544164.1 | |
| XM_011545864.1 | 4 | 530 | XP_011544166.1 | |
| AF037601.1 | 3 | 292 | AAF18267.1 | |
| AF151826.1 | 3 | 400 | AAD34063.1 | |
| AK025884.1 | 4 | 523 | BAB15270.1 | |
| AK223364.1 | 4 | 574 | BAD97084.1 | |
| AK225271.1 | 3 | 360 | ||
| AK291885.1 | 3 | 360 | ||
| AK314409.1 | 3 | 361 | ||
| AL137283.1 | 3 | 220 | CAB70677.1 | |
| AL560574.3 | 3 | 407 | ||
| BC001214.1 | 3 | 383 | AAH01214.1 | |
| BC014217.2 | 4 | 504 | AAH14217.1 | |
| BI459868.1 | 3 | 438 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758638 | Chr.16:24932074 - 25175657 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map