Assay Details
Target Gene Details
Entrez Gene ID: | 7421 |
Gene Name: | vitamin D (1,25- dihydroxyvitamin D3) receptor |
Gene Aliases: |
NR1I1, PPP1R163 |
Location: |
Chr.12:47841537-47905031 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VDR | NM_000376.2 | 10 | 1298 | NP_000367.1 |
| NM_001017535.1 | 11 | 1420 | NP_001017535.1 | |
| NM_001017536.1 | 10 | 1689 | NP_001017536.1 | |
| XM_006719587.3 | 9 | 1215 | XP_006719650.1 | |
| XM_011538720.2 | 10 | 1330 | XP_011537022.1 | |
| AB307700.1 | 8 | 1138 | ||
| AF026260.1 | 8 | 1138 | AAB95155.1 | |
| AK225844.1 | 10 | 1297 | ||
| AK299446.1 | 9 | 1362 | ||
| AK312267.1 | 10 | 1290 | ||
| BC060832.1 | 10 | 1661 | AAH60832.1 | |
| HQ692836.1 | 8 | 1138 | ||
| J03258.1 | 10 | 1253 | AAA61273.1 | |
| X67482.1 | 9 | 1188 | CAA47824.1 |
More Information
Set Membership: |
|

Genomic Map