Assay Details
Target Gene Details
Entrez Gene ID: | 3670 |
Gene Name: | ISL LIM homeobox 1 |
Gene Aliases: |
ISLET1, Isl-1 |
Location: |
Chr.5:51383124-51394730 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISL1 | NM_002202.2 | 6 | 1935 | NP_002193.2 |
| XM_011543380.2 | 5 | 3053 | XP_011541682.1 | |
| BC031213.1 | 6 | 1936 | AAH31213.1 | |
| U07559.1 | 6 | 1626 | AAA81946.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830288 | Chr.5:51302388 - 51459407 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map