Assay Details
Target Gene Details
Entrez Gene ID: | 2793 |
Gene Name: | G protein subunit gamma transducin 2 |
Gene Aliases: |
G-GAMMA-8, G-GAMMA-C, GNG8, GNG9, GNGT8 |
Location: |
Chr.17:49206234-49210574 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNGT2 | NM_001198754.1 | 4 | 854 | NP_001185683.1 |
| NM_001198755.1 | 4 | 881 | NP_001185684.1 | |
| NM_001198756.1 | 4 | 771 | NP_001185685.1 | |
| NM_031498.2 | 4 | 898 | NP_113686.1 | |
| AI857683.1 | 1 | 140 | ||
| BU742490.1 | 1 | 160 |
More Information
Set Membership: |
|

Genomic Map