Assay Details
Target Gene Details
Entrez Gene ID: | 25925 |
Gene Name: | zinc finger protein 521 |
Gene Aliases: |
EHZF, Evi3 |
Location: |
Chr.18:25061924-25352250 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF521 | NM_001308225.1 | 7 | 4806 | NP_001295154.1 |
| NM_015461.2 | 8 | 4847 | NP_056276.1 | |
| XM_011525909.2 | 8 | 4844 | XP_011524211.1 | |
| XM_011525911.1 | 9 | 4883 | XP_011524213.1 | |
| XM_017025697.1 | 7 | 4733 | XP_016881186.1 | |
| XM_017025698.1 | 9 | 4810 | XP_016881187.1 | |
| AF141339.1 | 5 | 1261 | AAG49442.1 | |
| AK027354.1 | 8 | 4749 | BAB55056.1 | |
| AK074046.1 | 5 | 4369 | ||
| CA415157.1 | 1 | 143 |
More Information
Set Membership: |
|

Genomic Map