Assay Details
Target Gene Details
Entrez Gene ID: | 9518 |
Gene Name: | growth differentiation factor 15 |
Gene Aliases: |
GDF-15, MIC-1, MIC1, NAG-1, PDF, PLAB, PTGFB |
Location: |
Chr.19:18386158-18389176 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDF15 | NM_004864.2 | 2 | 740 | NP_004855.2 |
| AB000584.1 | 2 | 740 | BAA19151.1 | |
| AF003934.1 | 2 | 742 | AAC24456.1 | |
| AF019770.1 | 2 | 725 | AAB88673.1 | |
| AK225431.1 | 2 | 740 | ||
| AK291530.1 | 2 | 740 | ||
| BC000529.2 | 2 | 719 | AAH00529.1 | |
| BC008962.2 | 2 | 725 | AAH08962.1 | |
| BT019465.1 | 2 | 708 | AAV38272.1 | |
| U51731.1 | 2 | 720 | ||
| U88323.1 | 2 | 733 | AAB88913.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952420 | Chr.19:18117791 - 18476990 on Build GRCh38 | Deletion |
|
| nsv2430 | Chr.19:18385618 - 18404127 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map