Assay Details
Target Gene Details
Entrez Gene ID: | 8732 |
Gene Name: | RNA guanylyltransferase and 5'-phosphatase |
Gene Aliases: |
CAP1A, HCE, HCE1, hCAP |
Location: |
Chr.6:88609897-88963716 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNGTT | NM_001286426.1 | 15 | 3390 | NP_001273355.1 |
| NM_001286428.1 | 14 | 3280 | NP_001273357.1 | |
| NM_003800.4 | 16 | 3459 | NP_003791.3 | |
| XM_017011401.1 | 17 | 3564 | XP_016866890.1 | |
| AF025654.1 | 16 | 3527 | AAB91559.1 | |
| BC019954.1 | 16 | 3426 | AAH19954.1 | |
| BX537450.1 | 3 | 4647 | CAD97693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6087n100 | Chr.6:88552751 - 88940524 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77260636] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map