Assay Details
Target Gene Details
Entrez Gene ID: | 9039 |
Gene Name: | ubiquitin like modifier activating enzyme 3 |
Gene Aliases: |
NAE2, UBE1C, hUBA3 |
Location: |
Chr.3:69054730-69080373 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBA3 | NM_003968.3 | 11 | 888 | NP_003959.3 |
| NM_198195.1 | 10 | 846 | NP_937838.1 | |
| XM_011534210.1 | 10 | 802 | XP_011532512.1 | |
| XM_011534211.1 | 9 | 755 | XP_011532513.1 | |
| AB012190.1 | 9 | 805 | BAA33144.1 | |
| AF046024.1 | 10 | 889 | AAC27648.1 | |
| AK002159.1 | 10 | 833 | ||
| AK289392.1 | 11 | 874 | ||
| AK298878.1 | 6 | 561 | ||
| AK314751.1 | 11 | 871 | ||
| AK316540.1 | 8 | 725 | ||
| AL117566.1 | 11 | 884 | CAB55996.2 | |
| BC022853.1 | 11 | 887 | AAH22853.1 | |
| CR533537.1 | 11 | 868 | CAG38568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520488 | Chr.3:69055601 - 69064854 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map