Assay Details
Target Gene Details
Entrez Gene ID: | 6490 |
Gene Name: | premelanosome protein |
Gene Aliases: |
D12S53E, ME20, ME20-M, ME20M, P1, P100, PMEL17, SI, SIL, SILV, gp100 |
Location: |
Chr.12:55954105-55966712 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PMEL | NM_001200053.1 | 4 | 638 | NP_001186982.1 |
| NM_001200054.1 | 6 | 896 | NP_001186983.1 | |
| NM_001320121.1 | 6 | 896 | NP_001307050.1 | |
| NM_001320122.1 | 6 | 896 | NP_001307051.1 | |
| NM_006928.4 | 7 | 931 | NP_008859.1 | |
| XM_006719569.1 | 6 | 1508 | XP_006719632.1 | |
| XM_011538685.1 | 7 | 960 | XP_011536987.1 | |
| AK092881.1 | 6 | 1509 | ||
| AK130180.1 | 7 | 955 | ||
| AK300150.1 | 4 | 618 | ||
| BC001414.2 | 6 | 855 | AAH01414.1 | |
| BT007202.1 | 6 | 845 | AAP35866.1 | |
| M32295.1 | 6 | 866 | AAA35930.1 | |
| M77348.1 | 6 | 856 | AAA60121.1 | |
| S73003.1 | 6 | 866 | AAC60634.1 | |
| U01874.1 | 6 | 851 | AAA18479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832424 | Chr.12:55881442 - 56073133 on Build GRCh38 | Loss |
|
| nsv1051961 | Chr.12:55868436 - 56235643 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map