Assay Details
Target Gene Details
Entrez Gene ID: | 10140 |
Gene Name: | transducer of ERBB2, 1 |
Gene Aliases: |
APRO5, APRO6, PIG49, TOB, TROB, TROB1 |
Location: |
Chr.17:50862223-50867978 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOB1 | NM_001243877.1 | 3 | 1048 | NP_001230806.1 |
| NM_001243885.1 | 2 | 859 | NP_001230814.1 | |
| NM_005749.3 | 2 | 1062 | NP_005740.1 | |
| AA675891.1 | ||||
| AK313904.1 | 3 | 1049 | ||
| AY524047.1 | 1 | 629 | AAS94256.1 | |
| BC031406.1 | 1 | 626 | AAH31406.1 | |
| BC070493.1 | 2 | 1052 | AAH70493.1 | |
| BC098415.1 | 2 | 1053 | AAH98415.1 | |
| BT019380.1 | 1 | 619 | AAV38187.1 | |
| BT019381.1 | 1 | 619 | AAV38188.1 | |
| CR536487.1 | 1 | 619 | CAG38726.1 | |
| CR541686.1 | 1 | 619 | CAG46487.1 | |
| D38305.1 | 1 | 663 | BAA07423.1 |
More Information
Set Membership: |
|

Genomic Map