Assay Details
Target Gene Details
Entrez Gene ID: | 5694 |
Gene Name: | proteasome subunit beta 6 |
Gene Aliases: |
DELTA, LMPY, Y |
Location: |
Chr.17:4796144-4798503 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMB6 | NM_001270481.1 | NP_001257410.1 | ||
| NM_002798.2 | NP_002789.1 | |||
| AK311768.1 | ||||
| BC000835.2 | AAH00835.1 | |||
| BF671774.1 | ||||
| BM553399.1 | ||||
| BQ691763.1 | ||||
| BT009809.1 | AAP88811.1 | |||
| BU848746.1 | ||||
| CR457065.1 | CAG33346.1 | |||
| D29012.1 | BAA06098.1 | |||
| X61971.1 | CAA43963.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833347 | Chr.17:4785297 - 4955981 on Build GRCh38 | Loss |
|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| nsv833346 | Chr.17:4646256 - 4860871 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map