Assay Details
Target Gene Details
Entrez Gene ID: | 127579 |
Gene Name: | DC-STAMP domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.1:155018513-155033781 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCST2 | NM_144622.2 | NP_653223.2 | ||
| XM_011509188.2 | XP_011507490.1 | |||
| XM_011509189.2 | 1 | 278 | XP_011507491.1 | |
| AK057496.1 | BAB71511.1 | |||
| AK097468.1 | BAC05065.1 | |||
| AK097993.1 | BAC05211.1 | |||
| BC111794.1 | AAI11795.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
| nsv952212 | Chr.1:154998225 - 155087624 on Build GRCh38 | Deletion |
|

Genomic Map