Assay Details
Target Gene Details
Entrez Gene ID: | 5293 |
Gene Name: | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta |
Gene Aliases: |
APDS, IMD14, P110DELTA, PI3K, p110D |
Location: |
Chr.1:9629889-9729114 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3CD | NM_005026.3 | NP_005017.3 | ||
| XM_006710689.2 | XP_006710752.1 | |||
| XM_017001477.1 | XP_016856966.1 | |||
| XM_017001478.1 | XP_016856967.1 | |||
| XM_017001479.1 | XP_016856968.1 | |||
| XM_017001480.1 | XP_016856969.1 | |||
| AK314191.1 | ||||
| BC150297.1 | ||||
| CN313981.1 | ||||
| U86453.1 | AAC25677.1 | |||
| Y10055.2 | CAA71149.2 |
Target Gene Details
Entrez Gene ID: | 644997 |
Gene Name: | PIK3CD antisense RNA 1 |
Gene Aliases: |
C1orf200 |
Location: |
Chr.1:9652610-9654586 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3CD-AS1 | NR_027045.1 | 2 | 854 | |
| AK097325.1 | 2 | 863 | BAC04998.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508847 | Chr.1:9612137 - 9678831 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map