Assay Details
Target Gene Details
Entrez Gene ID: | 2263 |
Gene Name: | fibroblast growth factor receptor 2 |
Gene Aliases: |
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25 |
Location: |
Chr.10:121478330-121598656 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGFR2 | NM_000141.4 | 18 | 4483 | NP_000132.3 |
| NM_001144914.1 | 15 | 3650 | NP_001138386.1 | |
| NM_001144915.1 | 17 | 3537 | NP_001138387.1 | |
| NM_001144916.1 | 15 | 3932 | NP_001138388.1 | |
| NM_001144917.1 | 16 | 4135 | NP_001138389.1 | |
| NM_001144918.1 | 16 | 4132 | NP_001138390.1 | |
| NM_001320654.1 | 17 | 3867 | NP_001307583.1 | |
| NM_001320658.1 | 18 | 4477 | NP_001307587.1 | |
| NM_022970.3 | 18 | 4486 | NP_075259.4 | |
| NM_023029.2 | 16 | 3719 | NP_075418.1 | |
| NR_073009.1 | 17 | 4286 | ||
| XM_006717708.2 | 18 | 4678 | XP_006717771.1 | |
| XM_006717710.3 | 18 | 4776 | XP_006717773.1 | |
| XM_006717711.2 | 16 | 3722 | XP_006717774.1 | |
| XM_006717712.2 | 15 | 3644 | XP_006717775.1 | |
| XM_017015920.1 | 18 | 4770 | XP_016871409.1 | |
| XM_017015921.1 | 18 | 4767 | XP_016871410.1 | |
| XM_017015922.1 | 16 | 3716 | XP_016871411.1 | |
| XM_017015923.1 | 16 | 3713 | XP_016871412.1 | |
| XM_017015924.1 | 16 | 4336 | XP_016871413.1 | |
| XM_017015925.1 | 16 | 4422 | XP_016871414.1 | |
| AB030073.1 | BAA89296.1 | |||
| CB305736.1 | 1 | 172 | ||
| M87770.1 | 18 | 4109 | AAA59470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428239 | Chr.10:120876200 - 121638984 on Build GRCh38 | Gain+Loss |
|
| nsv470972 | Chr.10:121148384 - 121482279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map