Assay Details
Target Gene Details
Entrez Gene ID: | 81570 |
Gene Name: | ClpB homolog, mitochondrial AAA ATPase chaperonin |
Gene Aliases: |
ANKCLB, HSP78, MEGCANN, MGCA7, SKD3 |
Location: |
Chr.11:72292425-72434684 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLPB | NM_001258392.2 | NP_001245321.1 | ||
| NM_001258393.2 | NP_001245322.1 | |||
| NM_001258394.2 | NP_001245323.1 | |||
| NM_030813.5 | NP_110440.1 | |||
| XM_005274320.1 | XP_005274377.1 | |||
| AK023214.1 | BAB14467.1 | |||
| AK302006.1 | ||||
| AK302069.1 | ||||
| AK302158.1 | ||||
| AL136909.1 | CAB66843.1 | |||
| AL834484.1 | CAD39142.1 | |||
| BC006257.2 | AAH06257.1 | |||
| BC006404.2 | AAH06404.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv385 | Chr.11:72270271 - 72306912 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map