Assay Details
Target Gene Details
Entrez Gene ID: | 8974 |
Gene Name: | prolyl 4-hydroxylase subunit alpha 2 |
Gene Aliases: |
- |
Location: |
Chr.5:132190147-132227863 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
More Information
Set Membership: |
|

Genomic Map