Assay Details
Target Gene Details
Entrez Gene ID: | 645432 |
Gene Name: | arrestin domain containing 5 |
Gene Aliases: |
- |
Location: |
Chr.19:4867614-4915469 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARRDC5 | XM_011528181.2 | XP_011526483.1 |
Target Gene Details
Entrez Gene ID: | 29128 |
Gene Name: | ubiquitin like with PHD and ring finger domains 1 |
Gene Aliases: |
ICBP90, Np95, RNF106, TDRD22, hNP95, hUHRF1, huNp95 |
Location: |
Chr.19:4903080-4962154 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UHRF1 | NM_001048201.2 | 1 | 52 | NP_001041666.1 |
| NM_001290050.1 | NP_001276979.1 | |||
| XM_011527942.2 | XP_011526244.1 | |||
| AB177623.1 | ||||
| AK074377.1 | ||||
| AK303361.1 | ||||
| AK307239.1 | ||||
| AK314579.1 | ||||
| DA451120.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065556 | Chr.19:4844193 - 4945238 on Build GRCh38 | Loss |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map