Assay Details
Target Gene Details
Entrez Gene ID: | 128869 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class U |
Gene Aliases: |
CDC91L1, GAB1 |
Location: |
Chr.20:34560542-34677285 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGU | NM_080476.4 | 11 | 1344 | NP_536724.1 |
| XM_011528542.2 | 5 | 635 | XP_011526844.1 | |
| XM_017027664.1 | 10 | 1018 | XP_016883153.1 | |
| AB086842.1 | 11 | 1150 | BAC53626.1 | |
| AK075507.1 | 11 | 1169 | BAC11660.1 | |
| AK301900.1 | 11 | 1144 | ||
| AY339061.1 | 10 | 1084 | AAQ18022.1 | |
| AY358816.1 | 11 | 1174 | AAQ89175.1 | |
| AY422169.1 | 11 | 1344 | AAR23798.1 | |
| BC030512.1 | 11 | 1155 | AAH30512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3359 | Chr.20:34531574 - 34576232 on Build GRCh38 | Deletion |
|
| nsv3360 | Chr.20:34562312 - 34597446 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map