Assay Details
Target Gene Details
Entrez Gene ID: | 11275 |
Gene Name: | kelch like family member 2 |
Gene Aliases: |
ABP-KELCH, MAV, MAYVEN |
Location: |
Chr.4:165207618-165323156 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL2 | NM_007246.3 | 1 | 244 | NP_009177.3 |
| XM_011531572.2 | 1 | 114 | XP_011529874.1 | |
| XM_011531575.2 | 1 | 188 | XP_011529877.1 | |
| XM_017007674.1 | 1 | 115 | XP_016863163.1 | |
| XM_017007676.1 | 1 | 174 | XP_016863165.1 | |
| XM_017007677.1 | 1 | 170 | XP_016863166.1 | |
| AF059569.1 | 1 | 92 | AAC67502.1 | |
| AK315372.1 | 1 | 89 | ||
| AL541403.3 | 1 | 244 | ||
| BC022503.1 | 1 | 226 | AAH22503.1 | |
| BC036468.2 | 1 | 185 | AAH36468.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26333 | Chr.4:165207285 - 165208055 on Build GRCh38 | Loss |
|
| nsv596050 | Chr.4:165207363 - 165208021 on Build GRCh38 | Gain+Loss |
|
| dgv2933n106 | Chr.4:165207448 - 165208148 on Build GRCh38 | Deletion |
|
| nsv1074235 | Chr.4:165207447 - 165207948 on Build GRCh38 | Deletion |
|
| nsv596048 | Chr.4:165206467 - 165207880 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map